missing translation for 'onlineSavingsMsg'
Learn More

Bromomethyl acetate, 95%

Product Code. 15403298
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15403298 5 g 5g
15413298 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15403298 Supplier Thermo Scientific Alfa Aesar Supplier No. H56755.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 590-97-6 | C3H5BrO2 | 152.98 g/mol

Bromomethyl acetate is involved in the preparation of optically active cyclohexene antisepsis agents. It acts as reagent in samarium diiodide-mediated conversion of ketones and aldehydes to 1,2-diacetates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromomethyl acetate is involved in the preparation of optically active cyclohexene antisepsis agents. It acts as reagent in samarium diiodide-mediated conversion of ketones and aldehydes to 1,2-diacetates.

Solubility
Miscible with chloroform.

Notes
Store in cool place. Moisture sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-97-6
Molecular Formula C3H5BrO2
Molecular Weight (g/mol) 152.98
MDL Number MFCD00000170
InChI Key NHYXMAKLBXBVEO-UHFFFAOYSA-N
Synonym bromomethanol acetate, methanol, bromo-, acetate, acetic acid bromomethyl ester, acetoxymethyl bromide, methanol, 1-bromo-, 1-acetate, brommethylacetat, acetoxymethylbromid, bromo-methyl acetate, acmc-20aljy, 1-bromomethyl acetate
PubChem CID 68536
IUPAC Name bromomethyl acetate
SMILES CC(=O)OCBr

Specifications

Density 1.56
Boiling Point 130°C to 133°C
Flash Point 57°C (135°F)
Linear Formula CH3CO2CH2Br
Quantity 5 g
UN Number UN3272
Sensitivity Moisture sensitive
Solubility Information Miscible with chloroform.
Formula Weight 152.98
Percent Purity 95%
Chemical Name or Material Bromomethyl acetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.