missing translation for 'onlineSavingsMsg'
Learn More

Bromomethyl acetate, 95%

Product Code. 10663015 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10663015 10 g Glass bottle 10g
10205054 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10663015 Supplier Thermo Scientific Acros Supplier No. 399180100
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 590-97-6 | C3H5BrO2 | 152.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-97-6
Molecular Formula C3H5BrO2
Molecular Weight (g/mol) 152.98
MDL Number MFCD00000170
InChI Key NHYXMAKLBXBVEO-UHFFFAOYSA-N
Synonym bromomethanol acetate, methanol, bromo-, acetate, acetic acid bromomethyl ester, acetoxymethyl bromide, methanol, 1-bromo-, 1-acetate, brommethylacetat, acetoxymethylbromid, bromo-methyl acetate, acmc-20aljy, 1-bromomethyl acetate
PubChem CID 68536
IUPAC Name bromomethyl acetate
SMILES CC(=O)OCBr

Specifications

Color White to Beige
Density 1.5600g/mL
Boiling Point 130.0°C to 133.0°C (750.0 mmHg)
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3CO2CH2Br
Refractive Index 1.4470 to 1.4540 (20°C, 589nm)
Quantity 10 g
Specific Gravity 1.56
Formula Weight 152.97
Percent Purity 95%
Physical Form Crystalline Powder or Powder
Chemical Name or Material Bromomethyl acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Bromomethyl acetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.