missing translation for 'onlineSavingsMsg'
Learn More

Bromomethyl Acetate (~90%), TRC

Product Code. 30133270
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133270 10 g 10g
30132691 5 g 5g
30012578 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133270 Supplier Toronto Research Chemicals Supplier No. TRCB68493010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-97-6
Molecular Formula C3 H5 Br O2
Molecular Weight (g/mol) 152.97
IUPAC Name bromomethyl acetate
SMILES CC(=O)OCBr

Specifications

Quantity 10 g
InChI Formula InChI=1S/C3H5BrO2/c1-3(5)6-2-4/h2H2,1H3
Formula Weight 151.9473
Chemical Name or Material Bromomethyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.