missing translation for 'onlineSavingsMsg'
Learn More

Bromomalonaldehyde, 97%

Product Code. 15454027
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15454027 50 g 50g
15434027 1 g 1g
15444027 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15454027 Supplier Thermo Scientific Alfa Aesar Supplier No. H31030.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2065-75-0 | C3H3BrO2 | 150.959 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2065-75-0
Molecular Formula C3H3BrO2
Molecular Weight (g/mol) 150.959
MDL Number MFCD00459999
InChI Key SURMYNZXHKLDFO-UHFFFAOYSA-N
Synonym 2-bromomalonaldehyde, bromomalonaldehyde, bromomalondialdehyde, propanedial, bromo, 2-bromomalondialdehyde, 2-bromo-1,3-propanedial, malonaldehyde, bromo, bromo-malonaldehyde, zlchem 220, 2-bromo-malonaldehyde
PubChem CID 74945
IUPAC Name 2-bromopropanedial
SMILES C(=O)C(C=O)Br

Specifications

Melting Point ∽145°C (decomposition)
Linear Formula BrCH(CHO)2
Quantity 50 g
Sensitivity Air Sensitive
Formula Weight 150.96
Percent Purity 97%
Chemical Name or Material Bromomalonaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.