missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromoform, TRC
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
Specifications
Specifications
| Chemical Name or Material | Tribromomethane |
| CAS | 75-25-2 |
| Molecular Formula | C H Br3 |
| Quantity | 25 mL |
| Synonym | Tribromomethane, Bromoform, Methenyl tribromide, NSC 8019 |
| InChI Formula | InChI=1S/CHBr3/c2-1(3)4/h1H |
| SMILES | BrC(Br)Br |
| IUPAC Name | bromoform |
| Molecular Weight (g/mol) | 252.73 |
| Formula Weight | 249.7628 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction