missing translation for 'onlineSavingsMsg'
Learn More

Bromoform, TRC

Product Code. 30344547
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 30344547

Brand: Toronto Research Chemicals TRCB687335100ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tribromomethane
CAS 75-25-2
Molecular Formula C H Br3
Quantity 100 mL
Synonym Tribromomethane, Bromoform, Methenyl tribromide, NSC 8019
InChI Formula InChI=1S/CHBr3/c2-1(3)4/h1H
SMILES BrC(Br)Br
IUPAC Name bromoform
Molecular Weight (g/mol) 252.73
Formula Weight 249.7628
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.