missing translation for 'onlineSavingsMsg'
Learn More

Bromoform, Technical, Unstabilised, SLR, Fisher Chemical

Product Code. 11493533 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493533 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11493533 Supplier Fisher Chemical Supplier No. B/4450/PB07
  • ONLINE EXCLUSIVE PRICE
    £222.00
    £207.00 / 250mL
    Save £15.00
    6% Off
 Restricted Item
Estimated Shipment: 16-04-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 75-25-2
Melting Point 8°C
Boiling Point 149°C
Packaging Plastic Coated Glass bottle
Molecular Formula CHBr3
MDL Number 128
Quantity 250 mL
Formula Weight 252.73g/mol
Physical Form Liquid
compliance-icons compliance-icons
Nombre del producto
  • Bromoform
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Indicaciones de peligro
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
Consejos de prudencia
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.