missing translation for 'onlineSavingsMsg'
Learn More

Bromoferrocene, 95%, Thermo Scientific Chemicals

Product Code. 11392019
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392019 5 g 5g
11382019 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392019 Supplier Thermo Scientific Alfa Aesar Supplier No. H26076.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Bromoferrocene is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromoferrocene is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1273-73-0
Molecular Formula C10H9BrFe
Molecular Weight (g/mol) 264.93
MDL Number MFCD03093913
IUPAC Name Bromoferrocene
SMILES [Fe].c1cccc1.Brc1cccc1

Specifications

Melting Point 31°C to 33°C
Quantity 5 g
Formula Weight 264.93
Percent Purity 95%
Chemical Name or Material Bromoferrocene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.