missing translation for 'onlineSavingsMsg'
Learn More

Bromodomain inhibitor-8, MedChemExpress

Product Code. 16486016
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486016 100 mg 100mg
16485418 10 mg 10mg
16428137 5 mg 5mg
16421607 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16486016 Supplier Medchem Express Supplier No. HY128703
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Bromodomain inhibitor-8 (Intermediate 21) is a BET bromodomain inhibitor for treating autoimmune and inflammatory diseases.

Bromodomain inhibitor-8 (Intermediate 21) is a BET bromodomain inhibitor for treating autoimmune and inflammatory diseases.

Specifications

Chemical Name or Material Bromodomain inhibitor-8
CAS 1300031-70-2
Color White
Purity Grade Notes Research
Molecular Formula C26H25ClN2O3
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (222.75 mM; Need ultrasonic)
SMILES O=C(OC)C1=CC=C(C2=CC3=C(N(C(C)=O)[C@@H](C)C[C@H]3NC4=CC=C(Cl)C=C4)C=C2)C=C1
Molecular Weight (g/mol) 448.94
Formula Weight 448.94
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.