missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Bromodichloromethane, 98%, stab. with potassium carbonate
Specifications
Specifications
| CAS | 75-27-4 |
| Melting Point | -57°C |
| Boiling Point | 87°C to 89°C |
| Odor | Chloroform-like |
| Molecular Formula | CHBrCl2 |
| MDL Number | MFCD00000824 |
| Quantity | 10 g |
| Beilstein | 1697005 |
| Merck Index | 14,1417 |
| Synonym | dichlorobromomethane, methane, bromodichloro, bromo dichloro methane, dichloromonobromomethane, monobromodichloromethane, bdcm, bromodichlormethane, unii-7ln464ch2o, ccris 819, stabilized with ethanol |
| Show More |
Safety and Handling
Product Identifier
- Bromotrichloromethane
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: Bromotrichloromethane
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?