missing translation for 'onlineSavingsMsg'
Learn More

Bromodeoxyuridine/BrdU Antibody (BRD494), Novus Biologicals™

Product Code. 18400862 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
Unit Size:
0.02mg
0.1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400862 0.02 mg 0.02mg
18410162 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18400862 Supplier Novus Biologicals Supplier No. NBP2329200.02mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Bromodeoxyuridine/BrdU Monoclonal antibody specifically detects Bromodeoxyuridine/BrdU in Nonspecies specific samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bromodeoxyuridine/BrdU
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BRD494
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5-1 ug/million cells, Immunohistochemistry 0.5-1 ug/mL, Immunocytochemistry/ Immunofluorescence 0.5-1 ug/mL, Immunohistochemistry-Paraffin 0.5-1.0ug/mL
Formulation 10 mM PBS with 0.05% BSA
Gene Alias Bromodeoxyuridine, BUdr
Host Species Mouse
Immunogen Bromodeoxyuridine/BrdU conjugated to KLH
Purification Method Protein A or G purified
Quantity 0.02 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.