missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromodeoxyuridine/BrdU Antibody (29G6.E8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-44055-0.05MG
This item is not returnable.
View return policy
Description
Bromodeoxyuridine/BrdU Monoclonal specifically detects Bromodeoxyuridine/BrdU in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Bromodeoxyuridine/BrdU | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000 - 1:5000, Flow Cytometry 1:50-1:100, ELISA 1:2000 - 1:10000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:500-1:3000 | |
| Bromodeoxyuridine, BUdr | |
| Anti-BrdU monoclonal antibody was produced in mice by repeated immunizations prepared via immunizations with BromodeoxyUridine-KLH followed by hybridoma development. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 29G6.E8 | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide | |
| Mouse | |
| 0.05 mg | |
| Non-species specific | |
| IgG1 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction