missing translation for 'onlineSavingsMsg'
Learn More

Bromocyclopentane, TRC

Product Code. 30333562
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30333562 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30333562 Supplier Toronto Research Chemicals Supplier No. TRCB68282550G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromocyclopentane
CAS 137-43-9
Molecular Formula C5H9Br
Quantity 50 g
Synonym Cyclopentyl Bromide, NSC 1110
InChI Formula InChI=1S/C5H9Br/c6-5-3-1-2-4-5/h5H,1-4H2
SMILES BrC1CCCC1
IUPAC Name bromocyclopentane
Molecular Weight (g/mol) 149.03
Formula Weight 147.9888
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.