missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Purple

Product Code. 11494097
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494097 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11494097 Supplier Thermo Scientific Alfa Aesar Supplier No. A19530.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 115-40-2 | C21H16Br2O5S | 540.22 g/mol

It is used as a pH indicator and to measure serum albumin concentrations. It is used as a pH indicator (5.2-6.8 : yellow to blue-purple) and also used in spectrophotometric determination of serum-albumin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pH indicator and to measure serum albumin concentrations. It is used as a pH indicator (5.2-6.8 : yellow to blue-purple) and also used in spectrophotometric determination of serum-albumin.

Solubility
Insoluble in water. Soluble in ethanol (80 mg/ml), alcohol, methanol (4 mg/ml), methyl cellosolve (300 mg/ml) and dilute alkalies.

Notes
Transition interval: pH 5.2 (yellow) to pH 6.8 (purple)
TRUSTED_SUSTAINABILITY

Specifications

Melting Point ∼240°C (decomposition)
CAS 115-40-2
Molecular Formula C21H16Br2O5S
MDL Number MFCD00011681
Beilstein 359618
Merck Index 14,1387
Solubility Information Insoluble aq. soln.. Soluble in ethanol (80 mg/ml),alcohol,methanol (4 mg/ml),methyl cellosolve (300 mg/ml) and dilute alkalies.
Quantity 250 g
InChI Key ABIUHPWEYMSGSR-UHFFFAOYSA-N
SMILES CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1
IUPAC Name 3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular Weight (g/mol) 540.22
PubChem CID 8273
ChEBI CHEBI:86154
Formula Weight 540.24
Chemical Name or Material Bromocresol Purple
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.