missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Purple, sodium salt, pure, water soluble, indicator

Product Code. 10581091 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
5g
This item is not returnable. View return policy

Product Code. 10581091

Brand: Thermo Scientific Chemicals 229610250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62625-30-3 | C21H15Br2NaO5S | 562.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62625-30-3
Molecular Formula C21H15Br2NaO5S
Molecular Weight (g/mol) 562.204
MDL Number MFCD00148896
InChI Key KVSJYIXUWNOWBD-UGAWPWHASA-M
Synonym BCP, Bromcresol Purple, sodium salt, 5';, 5';';-Dibromo-o-cresolsulfonephthalein, sodium salt
PubChem CID 23665568
IUPAC Name sodium;2-[(E)-(3-bromo-4-hydroxy-5-methylphenyl)-(3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
SMILES CC1=CC(=C(C2=CC=CC=C2S(=O)(=O)[O-])C3=CC(=C(C(=C3)Br)O)C)C=C(C1=O)Br.[Na+]

Specifications

Beilstein 19, 91
Merck Index 15, 1394
Solubility Information Solubility in water: Soluble
Infrared Spectrum Authentic
Formula Weight 562.2
Grade Pure
Packaging Glass bottle
Specific Extinction 900 min. (590nm, 0.01N NaOH)
Visual Transition Interval from pH 5.2 (yellow) to p 6.8 (purple)
Color Brown to Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Bromocresol Purple, sodium salt, Water soluble, Indicator, Pure
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.