missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Purple sodium salt, 0.04% w/v aq. soln.

Product Code. 11368967
Change view
Click to view available options
Quantity:
100 mL
500 mL
6 x 100 mL
Unit Size:
100mL
500mL
600mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368967 100 mL 100mL
11490151 500 mL 500mL
11378967 6 x 100 mL 600mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11368967 Supplier Thermo Scientific Alfa Aesar Supplier No. 038700.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62625-30-3 | C21H15Br2NaO5S | 562.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62625-30-3
Molecular Formula C21H15Br2NaO5S
Molecular Weight (g/mol) 562.204
MDL Number MFCD00148896
InChI Key KVSJYIXUWNOWBD-UGAWPWHASA-M
Synonym Bromocresol Purple, water soluble
PubChem CID 23665568
IUPAC Name sodium;2-[(E)-(3-bromo-4-hydroxy-5-methylphenyl)-(3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
SMILES CC1=CC(=C(C2=CC=CC=C2S(=O)(=O)[O-])C3=CC(=C(C(=C3)Br)O)C)C=C(C1=O)Br.[Na+]

Specifications

Concentration 0.04% w/v aq. soln.
Formula Weight 562.22
Color Orange
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Bromocresol Purple sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.