missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Green sodium salt

Product Code. 11428117
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428117 5 g 5g
11438117 25 g 25g
11448117 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428117 Supplier Thermo Scientific Chemicals Supplier No. A17503.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bromocresol Green sodium salt, C21H13Br4NaO5S, CAS Number-62625-32-5, Bromocresol Green, water soluble, 100g, 263-657-4, 720.01, ∼230 deg.C (decomposition), MFCD00148898, Ambient temperatures, Soluble in water and ethanol., Yes | CAS: 62625-32-5 | C21H13Br4NaO5S

Bromocresol Green sodium salt is used as a pH indicator. It is a pH sensitive triphenylmethane dye useful in various colorimetric detection studies and as an activity tracking dye for DNA agarose gel electrophoresis. It is also used in TLC (thin-layer chromatography) for visualizing compounds containing functional groups having pKa less than 5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromocresol Green sodium salt is used as a pH indicator. It is a pH sensitive triphenylmethane dye useful in various colorimetric detection studies and as an activity tracking dye for DNA agarose gel electrophoresis. It is also used in TLC (thin-layer chromatography) for visualizing compounds containing functional groups having pKa less than 5.

Solubility
Soluble in water and ethanol.

Notes
Transition interval: pH 3.8 (yellow) to pH 5.4 (blue)Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62625-32-5
Molecular Formula C21H13Br4NaO5S
MDL Number MFCD00148898
Synonym Bromocresol Green, water soluble

Specifications

Melting Point ∼230°C (decomposition)
Solubility Information Soluble in water and ethanol.
Formula Weight 720.01
Quantity 5 g
Chemical Name or Material Bromocresol Green sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.