missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green, Pure, Indicator Grade, Fisher Chemical™

Product Code. 10263180 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10263180 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10263180 Supplier Fisher Chemical Supplier No. B/4320/46

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 225°C
CAS 76-60-8
Molecular Formula C21H14Br4O5S
MDL Number 5874
Formula Weight 698.01g/mol
Packaging Glass Jar
Color Beige
Quantity 25 g
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.