missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Green, pure, indicator grade

Product Code. 10521211 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10521211 5 g Glass bottle 5g
10498870 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10521211 Supplier Thermo Scientific Chemicals Supplier No. 151310050
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Bromocresol Green, C21H14Br4O5S, CAS Number-76-60-8, 3', 3″, 5', 3',3″,5',5″-Tetrabromo-m-cresolsulfonephthalein, BCG, 5g, Brown, 200-972-8, 698.01, Pure, Warning, GHS H Statement: May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. | CAS: 76-60-8 | C21H14Br4O5S | 698.014 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-60-8
Molecular Formula C21H14Br4O5S
Molecular Weight (g/mol) 698.014
MDL Number MFCD00005874
InChI Key FRPHFZCDPYBUAU-UHFFFAOYSA-N
Synonym 3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein, BCG, 3', 3'', 5'
PubChem CID 6451
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br

Specifications

Melting Point 225°C, 437°F
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Extinction 600 min. (617nm, in buffer)
Visual Transition Interval from pH 3.8 (yellow) to pH 5.4 (blue)
Merck Index 15, 1393
Color Brown
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in benzene, ethanol (30g/L), very soluble in diethyl ether and ethyl acetate
Quantity 5 g
Physical Form Powder
Formula Weight 698.01
Grade Pure
Chemical Name or Material Bromocresol Green
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.