Learn More
Thermo Scientific Chemicals Bromocresol Green, ACS reagent
£16.80 - £56.80
Chemical Identifiers
CAS | 76-60-8 |
---|---|
Molecular Formula | C21H14Br4O5S |
Molecular Weight (g/mol) | 698.014 |
MDL Number | MFCD00005874 |
InChI Key | FRPHFZCDPYBUAU-UHFFFAOYSA-N |
Synonym | 3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein, BCG, 3', 3'', 5' |
PubChem CID | 6451 |
IUPAC Name | 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol |
SMILES | CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10453541
|
Thermo Scientific Chemicals
402930010 |
1 g | Glass bottle |
£16.80
1g |
Estimated Shipment: 27-09-2024
Log in to see stock. |
||||
10675861
|
Thermo Scientific Chemicals
402930050 |
5 g | Glass bottle |
£33.60
5g |
In Stock
Log in to see stock. |
||||
10740672
|
Thermo Scientific Chemicals
402930100 |
10 g | Glass bottle |
£56.80
10g |
Estimated Shipment: 27-09-2024
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
76-60-8 | |
698.014 | |
FRPHFZCDPYBUAU-UHFFFAOYSA-N | |
6451 | |
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br |
C21H14Br4O5S | |
MFCD00005874 | |
3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein, BCG, 3', 3'', 5' | |
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol |
Specifications
225.0°C | |
Passes Test | |
MFCD00005874 | |
3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein, BCG, 3', 3'', 5' | |
FRPHFZCDPYBUAU-UHFFFAOYSA-N | |
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br | |
698.014 | |
698.01 | |
from pH 3.8 (yellow) to pH 5.4 (blue) | |
Powder |
76-60-8 | |
C21H14Br4O5S | |
15, 1393 | |
Solubility in water: slightly soluble. Other solubilities: soluble in benzene, ethanol (30g/L), very soluble in diethyl ether and ethylacetate | |
Authentic | |
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol | |
6451 | |
ACS Reagent | |
Beige to Brown | |
Bromocresol Green |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
missing translation for 'einecsNumber' : 200-972-8
missing translation for 'rtecsNumber' : SJ7456000
missing translation for 'tsca' : TSCA
RUO – Research Use Only