missing translation for 'onlineSavingsMsg'
Learn More

Bromochlorophenol Blue, sodium salt, pure, water soluble, Thermo Scientific™

Product Code. 10012430 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10012430

Brand: Thermo Scientific Acros 199680050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 102185-52-4
Molecular Formula C19H9Br2Cl2O5S-
MDL Number MFCD00001612
InChI Key KGDDIJKLMLNSCH-LICLKQGHSA-M
SMILES C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Cl)C3=CC(=C(C(=C3)Br)O)Cl)S(=O)(=O)[O-]
IUPAC Name 2-[(Z)-(3-bromo-5-chloro-4-hydroxyphenyl)-(3-bromo-5-chloro-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
Molecular Weight (g/mol) 580.044
PubChem CID 131698806
Formula Weight 603.03
Grade Pure
Packaging Glass bottle
Quantity 5 g
Chemical Name or Material Bromochlorophenol Blue, sodium salt
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.