missing translation for 'onlineSavingsMsg'
Learn More

Bromochloronitromethane (>85%), TRC

Product Code. 30134873
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134873 2.5 mg 2.5mg
30137384 5 mg 5mg
17984095 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134873 Supplier Toronto Research Chemicals Supplier No. TRCB6823652.5MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135531-25-8
Molecular Formula C H Br Cl N O2
Molecular Weight (g/mol) 174.38
Synonym Bromochloronitromethane
IUPAC Name bromo-chloro-nitromethane
SMILES [O-][N+](=O)C(Cl)Br

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/CHBrClNO2/c2-1(3)4(5)6/h1H
Formula Weight 172.8879
Chemical Name or Material Bromochloronitromethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.