Learn More
R&D Systems™ BromoCatch™ GFP Expression Plasmid (C-term, pTK, Puro)
Product Code. 31900331
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Product Code. Quantity Unit Size
31900331 50 μg 50µg
1 options
Product Code. 31900331 Supplier R&D Systems™ Supplier No. RDELBC02GFP050
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This plasmid enables expression of proteins fused to the BromoCatch™ tag, a covalent self-labeling tag derived from a BET bromodomain.
TRUSTED_SUSTAINABILITY
Specifications
Product Line R&D Systems
Quantity 50 μg