missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromobenzene-d5, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Bromobenzene-D5 |
| CAS | 4165-57-5 |
| Molecular Formula | C6 2H5 Br |
| Quantity | 1 g |
| Synonym | Benzene-1,2,3,4,5-d5, 6-bromo- (ACI), 6-Bromobenzene-1,2,3,4,5-d5 (ACI), Benzene-d5, bromo- (6CI, 7CI, 8CI, 9CI), 1-Bromobenzene-2,3,4,5,6-d5, Bromobenzene-d5, Bromopentadeuteriobenzene, Bromopentadeuterobenzene, d5-Bromobenzene, Pentadeutero-1-bromobenzene, Pentadeuterophenyl bromide, Perdeuteriobromobenzene, Bromobenzene-D5 |
| InChI Formula | InChI=1S/C6H5Br/c7-6-4-2-1-3-5-6/h1-5H/i1D,2D,3D,4D,5D |
| SMILES | [2H]c1c([2H])c([2H])c(Br)c([2H])c1[2H] |
| IUPAC Name | 1-bromo-2,3,4,5,6-pentadeuteriobenzene |
| Molecular Weight (g/mol) | 162.04 |
| Formula Weight | 160.9888 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction