missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetyl chloride, 95%

Product Code. 10749585 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10749585

Brand: Thermo Scientific Acros 197991000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22118-09-8 | C2H2BrClO | 157.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22118-09-8
Molecular Formula C2H2BrClO
Molecular Weight (g/mol) 157.39
MDL Number MFCD00000724
InChI Key SYZRZLUNWVNNNV-UHFFFAOYSA-N
Synonym bromoacetyl chloride, acetyl chloride, bromo, bromo-acetyl chloride, monobromoacetic acid chloride, bromoacetylchloride, bromacetylchioride, bromacetylchloride, bromoacetylchioride, bromacetyl chloride, bromo-acetylchloride
PubChem CID 89602
IUPAC Name 2-bromoacetyl chloride
SMILES C(C(=O)Cl)Br

Specifications

Color Yellow to Brown
Density 1.9000g/mL
Boiling Point 127.0°C to 128.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2COCl
Refractive Index 1.496
Quantity 100 g
Beilstein 02,215
Specific Gravity 1.9
Solubility Information Solubility in water: reacts
Formula Weight 157.39
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Bromoacetyl chloride
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.