missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetonitrile, Dr. Ehrenstorfer

Product Code. 30321968
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
30321968 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 30321968

Brand: Dr. Ehrenstorfer DRECA10699000

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromoacetonitrile
CAS 590-17-0
Analyte or Component Names 2-Bromoacetonitrile
Shipping Condition +20°C
Molecular Formula C2H2BrN
Certifications/Compliance ISO/IEC 17025
Synonym Bromoacetonitrile, Bromomethyl Cyanide, Cyanomethyl Bromide, Acetonitrile, bromo- (6CI,7CI,8CI,9CI), 2-Bromoacetonitrile, Bromoacetonitrile, Bromomethyl cyanide, Cyanomethyl bromide
SMILES BrCC#N
Molecular Weight (g/mol) 119.95
Quantity 250 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.