missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetic anhydride, 95%

Product Code. p-4089774 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10244884

Brand: Thermo Scientific Acros 426700050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13094-51-4 | C4H4Br2O3 | 259.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13094-51-4
Molecular Formula C4H4Br2O3
Molecular Weight (g/mol) 259.87
MDL Number MFCD00045041
InChI Key FUKOTTQGWQVMQB-UHFFFAOYSA-N
Synonym bromoacetic anhydride, 2-bromoacetic anhydride, acetic acid, 2-bromo-,1,1'-anhydride, 2-bromoacetyl 2-bromoacetate, bromoaceticanhydride, acetic acid, bromo-, anhydride
PubChem CID 4259817
IUPAC Name (2-bromoacetyl) 2-bromoacetate
SMILES C(C(=O)OC(=O)CBr)Br

Specifications

Melting Point 40°C to 44°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (ex Br) (Argentometry)
Packaging Glass bottle
Linear Formula BrCH2C(=O)OC(=O)CH2Br
Quantity 5 g
Formula Weight 259.87
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Bromoacetic anhydride
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.