missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetaldehyde dimethyl acetal, 97%, stabilized

Product Code. 10781525 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10781525 25 mL Glass bottle 25mL
10042840 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10781525 Supplier Thermo Scientific Acros Supplier No. 221870250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7252-83-7 | C4H9BrO2 | 169.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7252-83-7
Molecular Formula C4H9BrO2
Molecular Weight (g/mol) 169.02
MDL Number MFCD00000213
InChI Key FUSFWUFSEJXMRQ-UHFFFAOYSA-N
Synonym bromoacetaldehyde dimethyl acetal, ethane, 2-bromo-1,1-dimethoxy, 2,2-dimethoxyethyl bromide, dimethylbromoacetal, 1-bromo-2,2-dimethoxyethane, 2,2-dimethoxybromoethane, 2-bromoacetaldehyde dimethyl acetal, bromoacetoaldehyde dimethyl acetal, bromoacetaldehyde dimethylacetal, acetaldehyde, bromo-, dimethyl acetal
PubChem CID 81672
IUPAC Name 2-bromo-1,1-dimethoxyethane
SMILES COC(CBr)OC

Specifications

Density 1.4300g/mL
Boiling Point 148.0°C to 150.0°C
Flash Point 51°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2CH(OCH3)2
Refractive Index 1.4450 to 1.4470
Quantity 25 mL
Beilstein 01, 624
Specific Gravity 1.43
Solubility Information Solubility in water: insoluble
Formula Weight 169.02
Percent Purity 97%
Chemical Name or Material Bromoacetaldehyde dimethyl acetal, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Bromoacetaldehyde dimethyl acetal, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H301-Toxic if swallowed.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.