missing translation for 'onlineSavingsMsg'
Learn More

Bromo-tris-pyrrolidino-phosphonium hexafluorophosphate, 97%

Product Code. 10339210 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10339210 5 g Glass bottle 5g
10237700 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10339210 Supplier Thermo Scientific Acros Supplier No. 302130050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 132705-51-2 | C12H24BrN3P·F6P | 466.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132705-51-2
Molecular Formula C12H24BrN3P·F6P
Molecular Weight (g/mol) 466.19
MDL Number MFCD00077412
InChI Key CYKRMWNZYOIJCH-UHFFFAOYSA-N
Synonym pybrop, bromo-tris-pyrrolidino-phosphonium hexafluorophosphate, bromotripyrrolidinophosphonium hexafluorophosphate, bromotri pyrrolidin-1-yl phosphonium hexafluorophosphate v, pybrop r, bromotri 1-pyrrolidinyl phosphonium hexafluorophosphate, bromotri pyrrolidino phosphonium hexafluorophosphate, bromo-tris-pyrrolidinophosphoniumhexafluorophosphate, bromo-tris-pyrrolidino phosphoniumhexafluorophosphate, bromo-tris-pyrrolidino-phosphoniumhexafluorophosphate
PubChem CID 2733179
IUPAC Name bromo(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate
SMILES C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)Br.F[P-](F)(F)(F)(F)F

Specifications

Melting Point >100°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: moderately soluble in water
Formula Weight 466.19
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Bromo-tris-pyrrolidino-phosphonium hexafluorophosphate, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.