missing translation for 'onlineSavingsMsg'
Learn More

α'-Bromo-α,α,α-trifluoro-m-xylene, 98%, ACROS Organics™

Product Code. 10553041 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10553041 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10553041 Supplier Thermo Scientific Acros Supplier No. 312980050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 402-23-3
CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C8H6BrF3
Linear Formula CF3C6H4CH2Br
Refractive Index 1.4910 to 1.4930 (20°C, 589nm)
MDL Number MFCD00000395
Quantity 5 g
Synonym 3-trifluoromethyl benzyl bromide, 1-bromomethyl-3-trifluoromethyl benzene, m-trifluoromethylbenzyl bromide, 3-trifluoromethylbenzyl bromide, 3-bromomethyl benzotrifluoride, 3-trifluoromethylbenzylbromide, 3-trifluoromethyl benzylbromide, 1-bromomethyl-3-trifluoromethyl-benzene, alpha'-bromo-alpha,alpha,alpha-trifluoro-m-xylene
InChI Key MYYYZNVAUZVXBO-UHFFFAOYSA-N
SMILES FC(F)(F)C1=CC=CC(CBr)=C1
IUPAC Name 1-(bromomethyl)-3-(trifluoromethyl)benzene
Molecular Weight (g/mol) 239.04
PubChem CID 123061
Formula Weight 239.04
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material α'-Bromo-α, α, α-trifluoro-m-xylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.