missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-p-xylene, 98%

Product Code. 10085943 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10085943 100 g Glass bottle 100g
10733203 5 g Glass bottle 5g
10746131 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10085943 Supplier Thermo Scientific Supplier No. 107571000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 104-81-4 | C8H9Br | 185.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-81-4
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.06
MDL Number MFCD00000181
InChI Key WZRKSPFYXUXINF-UHFFFAOYSA-N
Synonym 4-methylbenzyl bromide, 1-bromomethyl-4-methylbenzene, p-xylyl bromide, p-methylbenzyl bromide, alpha-bromo-p-xylene, benzene, 1-bromomethyl-4-methyl, 4-bromomethyl toluene, p-bromomethyl toluene, alpha-bromo-p-xylol, 1-bromomethyl-4-methyl-benzene
PubChem CID 7721
IUPAC Name 1-(bromomethyl)-4-methylbenzene
SMILES CC1=CC=C(C=C1)CBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 33.0°C to 37.0°C
Color White to Yellow
Boiling Point 218.0°C to 220.0°C
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CH2Br
Quantity 100 g
Beilstein 05, 385
Merck Index 15, 10288
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in chloroform and ether
Formula Weight 185.06
Percent Purity 98%
Physical Form Crystalline Mass
Chemical Name or Material α-Bromo-p-xylene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • a-Bromo-p-xylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.