missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-o-tolunitrile, 98%

Product Code. 10008571 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10008571 5 g Glass bottle 5g
10554101 25 g Glass bottle 25g
10205740 100 g Glass bottle 100g
10761614 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10008571 Supplier Thermo Scientific Acros Supplier No. 163130050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22115-41-9 | C8H6BrN | 196.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22115-41-9
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.05
MDL Number MFCD00001794
InChI Key QGXNHCXKWFNKCG-UHFFFAOYSA-N
Synonym 2-bromomethyl benzonitrile, 2-cyanobenzyl bromide, alpha-bromo-o-tolunitrile, o-cyanobenzyl bromide, a-bromo-o-tolunitrile, alpha bromo-o-tolunitrile, 2 bromomethyl benzonitrile, 2-bromomethyl-benzonitrile, 2-cyanophenyl methyl bromide, benzonitrile, 2-bromomethyl
PubChem CID 89599
IUPAC Name 2-(bromomethyl)benzonitrile
SMILES BrCC1=CC=CC=C1C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 70°C to 74°C
Color Brown-Gray to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2C6H4CN
Quantity 5 g
Beilstein 09, 470
Formula Weight 196.05
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material α-Bromo-o-tolunitrile
Show More Show Less
compliance-icons
Product Identifier
  • a-Bromo-o-tolunitrile
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.