missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-2'-acetonaphthone, 98%, Thermo Scientific Chemicals

Catalog Number 10346740
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
613-54-7
MFCD00004109
YHXHHGDUANVQHE-UHFFFAOYSA-N
2-bromo-2'-acetonaphthone, bromomethyl 2-naphthyl ketone, 2-bromoacetyl naphthalene, 2-bromo-1-naphthalen-2-yl ethanone, 2-bromo-1-2-naphthyl-1-ethanone, ethanone, 2-bromo-1-2-naphthalenyl, 2-bromo-1-2-naphthyl ethanone, 2-bromo-1-naphthalen-2-yl ethan-1-one, alpha-bromo-2'-acetonaphthone, 2-bromo-1-naphthalen-2-yl-ethanone
69179
2-bromo-1-naphthalen-2-ylethanone
C1=CC=C2C=C(C=CC2=C1)C(=O)CBr
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
613-54-7
MFCD00004109
YHXHHGDUANVQHE-UHFFFAOYSA-N
2-bromo-2'-acetonaphthone, bromomethyl 2-naphthyl ketone, 2-bromoacetyl naphthalene, 2-bromo-1-naphthalen-2-yl ethanone, 2-bromo-1-2-naphthyl-1-ethanone, ethanone, 2-bromo-1-2-naphthalenyl, 2-bromo-1-2-naphthyl ethanone, 2-bromo-1-naphthalen-2-yl ethan-1-one, alpha-bromo-2'-acetonaphthone, 2-bromo-1-naphthalen-2-yl-ethanone
69179
2-bromo-1-naphthalen-2-ylethanone
C1=CC=C2C=C(C=CC2=C1)C(=O)CBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 82°C to 85°C
Color Brown-Purple
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C12H9BrO
Linear Formula C10H7COCH2Br
Quantity 10 g
Molecular Weight (g/mol) 249.11
Formula Weight 249.11
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material α-Bromo-2'-acetonaphthone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • a-Bromo-2'-acetonaphthone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'