Product Code. 10338970
Shop All Thermo Scientific Chemicals Products
Click to view available options
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10338970
5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
10338970
Supplier
Thermo Scientific Acros
Supplier No.
303580050
This item is currently unavailable or has been discontinued. View the product page for possible alternatives.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Specifications
CAS
23915-07-3
CAS Min %
97.5
CAS Max %
100.0
Color
Yellow
Density
1.6100g/mL
Flash Point
39°C
Infrared Spectrum
Authentic
Assay Percent Range
98%
Packaging
Glass bottle
Molecular Formula
C7H5BrF2
Linear Formula
F2 C6 H3 CH2 Br
Refractive Index
1.5240 to 1.5260
MDL Number
MFCD00011649
Quantity
5 g
Synonym
2,4-difluorobenzyl bromide, 1-bromomethyl-2,4-difluorobenzene, 2,4-difluorobenzylbromide, alpha-bromo-2,4-difluorotoluene, benzene, 1-bromomethyl-2,4-difluoro, 2,4-difluoro benzyl bromide, 1-bromomethyl-2,4-difluoro-benzene, 2-chloromethyl quinoline hcl salt, fr cf d1e, pubchem4911
InChI Key
IBLMYGXJKQIGSN-UHFFFAOYSA-N
SMILES
C1=CC(=C(C=C1F)F)CBr
IUPAC Name
1-(bromomethyl)-2,4-difluorobenzene
Molecular Weight (g/mol)
207.018
PubChem CID
90297
Formula Weight
207.02
Percent Purity
98%
Physical Form
Liquid
Chemical Name or Material
α-Bromo-2, 4-difluorotoluene
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?