missing translation for 'onlineSavingsMsg'
Learn More

Bromo-2,3,4-tri-O-benzoyl-Alpha-D-glucuronic Acid Methyl, TRC

Product Code. 30345781
Ansicht ändern
Click to view available options
Quantity:
100 mg
Packungsgröße:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
30345781 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30345781 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCB688353100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromo-2,3,4-tri-O-benzoyl-a-D-glucuronic Acid Methyl Ester
CAS 103674-69-7
Molecular Formula C28 H23 Br O9
Quantity 100 mg
Synonym 2,3,4-Tri-O-Benzoyl-5-Bromo-α-D-Glucuronic Acid Methyl Ester
InChI Formula InChI=1S/C28H23BrO9/c1-34-28(33)23-21(37-26(31)18-13-7-3-8-14-18)20(36-25(30)17-11-5-2-6-12-17)22(24(29)35-23)38-27(32)19-15-9-4-10-16-19/h2-16,20-24H,1H3/t20-,21-,22+,23-,24-/m0/s1
SMILES COC(=O)[C@H]1O[C@H](Br)[C@H](OC(=O)c2ccccc2)[C@@H](OC(=O)c3ccccc3)[C@@H]1OC(=O)c4ccccc4
IUPAC Name methyl (2S,3S,4S,5R,6R)-3,4,5-tribenzoyloxy-6-bromooxane-2-carboxylate
Molecular Weight (g/mol) 583.38
Formula Weight 582.0525
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.