missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromo 2,3,4,6-Tetra-O-acetyl-Alpha-D-glucopyranoside (~ 5% CaCO3 as stabilizer), TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Bromo 2,3,4,6-Tetra-O-acetyl-a-D-glucopyranoside |
| CAS | 572-09-8 |
| Molecular Formula | C14 H19 Br O9 |
| Quantity | 25 g |
| Synonym | Acetobromo-D-glucose |
| InChI Formula | InChI=1S/C14H19BrO9/c1-6(16)20-5-10-11(21-7(2)17)12(22-8(3)18)13(14(15)24-10)23-9(4)19/h10-14H,5H2,1-4H3/t10-,11-,12+,13-,14+/m1/s1 |
| SMILES | CC(=O)OC[C@H]1O[C@H](Br)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C |
| IUPAC Name | [(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxan-2-yl]methyl acetate |
| Molecular Weight (g/mol) | 411.2 |
| Formula Weight | 410.0212 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction