missing translation for 'onlineSavingsMsg'
Learn More

Bromisoval, MedChemExpress

Product Code. 16472369
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16472369 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16472369 Supplier Medchem Express Supplier No. HYB2113
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Bromisoval has anti-inflammatory effects.

Bromisoval has anti-inflammatory effects.

Specifications

Chemical Name or Material Bromisoval
CAS 496-67-3
Color White
Purity Grade Notes Research
Molecular Formula C6H11BrN2O2
Quantity 1 g
Synonym Bromovalerylurea
Solubility Information DMSO : 300 mg/mL (1344.87 mM; Need ultrasonic and warming)
SMILES CC(C)C(Br)C(NC(N)=O)=O
Molecular Weight (g/mol) 223.07
Formula Weight 223.07
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.