missing translation for 'onlineSavingsMsg'
Learn More

Bromine liquid, 99.8%, Thermo Scientific Chemicals

Product Code. 15410977
Click to view available options
Quantity:
25 g
250 g
1 kg
5 x 1 kg

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7726-95-6
Molecular Formula Br
Molecular Weight (g/mol) 79.91
MDL Number MFCD00010896
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
Synonym bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
PubChem CID 24408
ChEBI CHEBI:29224
SMILES [Br-]

Specifications

Physical Form Liquid
Quantity 250 g
Assay Percent Range 99.8%
IUPAC Name bromide
Formula Weight 159.81
Percent Purity 99.8%
Density 3.1g/cm3
Chemical Name or Material Bromine, liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bromine liquid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : bromine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'