missing translation for 'onlineSavingsMsg'
Learn More

Bromine liquid, 99.8%

Product Code. p-3256937
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
5 x 1 kg
Unit Size:
1kg
250g
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15400977 25 g 25g
15410977 250 g 250g
15420977 1 kg 1kg
15430977 5 x 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15400977 Supplier Thermo Scientific Alfa Aesar Supplier No. 000905.14
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7726-95-6 | Br | 79.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7726-95-6
Molecular Formula Br
Molecular Weight (g/mol) 79.91
MDL Number MFCD00010896
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
Synonym bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
PubChem CID 24408
ChEBI CHEBI:29224
SMILES [Br-]

Specifications

Physical Form Liquid
Quantity 25 g
Assay Percent Range 99.8%
IUPAC Name bromide
Formula Weight 159.81
Percent Purity 99.8%
Density 3.1g/cm3
Chemical Name or Material Bromine
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bromine liquid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Acute toxicity Category 2
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P320-Specific treatment is urgent (see on this label).
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.