missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromine concentrate, For 1L standard solution, 0.05 M Br2, Solstice
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| CAS Min % | ≥70.0000%, ≥10.0000% |
| CAS Max % | ≤90.0000%, ≤30.0000% |
| Physical Form | Concentrate |
| Quantity | 6 x 1 Ea. |
| Linear Formula | BaCl2 |
| MDL Number | MFCD00003445 |
| Packaging | Ampoule |
| Density | 1.307 g/mL at 25 °C(lit.) |
| Chemical Name or Material | Bromine concentrate, For 1L standard solution, 0.05 M Br2 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?