missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromindione, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Bromindione |
| CAS | 1146-98-1 |
| Molecular Formula | C15 H9 Br O2 |
| Quantity | 100 mg |
| Synonym | 1H-Indene-1,3(2H)-dione, 2-(4-bromophenyl)-, 1,3-Indandione, 2-(p-bromophenyl)- (6CI,7CI,8CI), 2-(4-Bromophenyl)-1H-indene-1,3(2H)-dione, 2-(4-Bromophenyl)-1,3-indandione, 2-(p-Bromophenyl)-1,3-indandione, Bromindione, Bromophendione, Bromophenindione, Circladin, Fluidane, Fluidemin, Fluidemol, HL 255, Haldinone, Halinone, M. G. 2555 |
| InChI Formula | InChI=1S/C15H9BrO2/c16-10-7-5-9(6-8-10)13-14(17)11-3-1-2-4-12(11)15(13)18/h1-8,13H |
| SMILES | Brc1ccc(cc1)C2C(=O)c3ccccc3C2=O |
| IUPAC Name | 2-(4-bromophenyl)indene-1,3-dione |
| Molecular Weight (g/mol) | 301.13 |
| Formula Weight | 299.9786 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction