missing translation for 'onlineSavingsMsg'
Learn More

Bromide (from NH4Br) Standard: Br- @ 10000 μg/mL in H2O, VHG™

Product Code. 30003097
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 30003097

Brand: VHG Labs VHGI1PABR500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Bromide (from NH4Br) Standard: Br- @ 10000 μg/mL in H2O
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromide
CAS 24959-67-9
Methods Ion Chromatography/Wet Chemistry
Analyte or Component Names Bromide
Shipping Condition Room Temperature
Molecular Formula Br-
Certifications/Compliance ISO 17034
Synonym Br−
SMILES [Br-]
IUPAC Name Bromide
Solvent or Matrix Water
Molecular Weight (g/mol) 79.9
Quantity 500 mL
Formula Weight 79.9 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.