missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromfenac sodium hydrate, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
Description
Bromfenac sodium hydrate (Bromfenac monosodium salt sesquihydrate) is a potent and orally active inhibitor of COX, with IC50s of 5.56 and 7.45 nM for COX-1 and COX-2, respectively. Bromfenac sodium hydrate can be used in ocular inflammation research.
Specifications
Specifications
| Chemical Name or Material | Bromfenac sodium hydrate |
| CAS | 120638-55-3 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C15H14BrNNaO4.5 |
| Quantity | 50 mg |
| Synonym | Bromfenac monosodium salt sesquihydrate |
| Solubility Information | DMSO : ≥ 100 mg/mL (260.98 mM) ∣H2O : ≥ 100 mg/mL (260.98 mM) |
| SMILES | O=C(O[Na])CC1=CC=CC(C(C2=CC=C(Br)C=C2)=O)=C1N.[1.5H2O] |
| Molecular Weight (g/mol) | 383.17 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction