missing translation for 'onlineSavingsMsg'
Learn More

Oxoid Brochothrix thermosphacta ATCC™

Product Code. 12995903
Change view
Click to view available options
Quantity:
5 Pk.
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12995903 5 Pk. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
If you would like to order, a signed ATCC agreement is required.
Product Code. 12995903 Supplier Oxoid Supplier No. R4609038
  • ONLINE EXCLUSIVE PRICE
    £249.00
    £226.03 / Pack of 5
    Save £22.97
    9% Off
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
If you would like to order, a signed ATCC agreement is required.

Brochothrix thermosphacta ATCC™ from Oxoid is a gram positive rod.

  • Gram-positive rod
  • Constitutes significant proportion of spoilage flora of meat and meat products
  • Not considered to be pathogenic, it is economically important meat-spoilage organism
    • Grows in wide variety of meats and meat products
    • Produces malodorous metabolic end products, make affected meat unpalatable
  • STAA medium made selective by inclusion of streptomycin sulphate, thallous acetate and actidione (cycloheximide)
  • Medium modified from original formulation by removal of toxic selective agent cycloheximide as recommended by NMKL
  • According to this method this medium is applicable for meat, meat products and some fish products
TRUSTED_SUSTAINABILITY

Specifications

Biosafety Level 1
Form Freeze-dried
For Use With (Application) Produces restriction endonuclease MthI; Bacteriophage host
No. per Pack 5
Quantity 5 Pk.
Species Brochothrix thermosphacta
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.