missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRN4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92361-0.02ml
This item is not returnable.
View return policy
Description
BRN4 Polyclonal antibody specifically detects BRN4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| BRN4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| brain-specific homeobox/POU domain protein 4, octamer-binding transcription factor 9, OTF9, POU class 3 homeobox 4, POU domain, class 3, transcription factor 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human POU3F4 (NP_000298.3). MATAASNPYSILSSTSLVHADSAGMQQGSPFRNPQKLLQSDYLQGVPSNGHPLGHHWVTSLSDGGPWSSTLATSPLDQQDVKPGREDLQLGAIIHHRSPHVAHHSPHTNHPNAWGASPAPNPSITSSGQPLNVYSQPGFTVSGMLEHGGLTPPPAAASAQSLHPVLREPPDHGELGSHHCQDHSDEETPT | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5456 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction