missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRN3B/POU4F2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35901-20ul
This item is not returnable.
View return policy
Description
BRN3B/POU4F2 Polyclonal antibody specifically detects BRN3B/POU4F2 in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| BRN3B/POU4F2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| POU domain, class 4, transcription factor 2, BRN3.2, Brn-3b, BRN3B, POU class 4 homeobox 2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human BRN3B/POU4F2 (NP_004566.2).,, Sequence:, MMMMSLNSKQAFSMPHGGSLHVEPKYSALHSTSPGSSAPIAPSASSPSSSSNAGGGGGGGGGGGGGGGRSSSSSSSGSSGGGGSEAMRRACLPTPPSNIF | |
| 20 μL | |
| Neuroscience | |
| 5458 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction