missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BRMS1L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532016
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (93%), Chicken (99%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene shows sequence similarity to the human breast carcinoma metastasis suppressor (BRMS1) protein and the mammalian Sds3 (suppressor of defective silencing 3) proteins. This protein is a component of the mSin3a family of histone deacetylase complexes (HDAC).
Specifications
| BRMS1L | |
| Polyclonal | |
| Unconjugated | |
| BRMS1L | |
| 0710008O11Rik; AI159718; breast cancer metastasis-suppressor 1; breast cancer metastasis-suppressor 1-like; breast cancer metastasis-suppressor 1-like protein; BRMS1; BRMS1-homolog protein p40; BRMS1L; BRMS1-like protein p40; D12Ertd407e | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84312 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q5PSV4 | |
| BRMS1L | |
| Recombinant protein encompassing a sequence within the center region of human BRMS1L. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction