missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BRMS1L Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561262
This item is not returnable.
View return policy
Description
Immunogen sequence: EDWTTIRKAM ATLGPHRVKT EPPVKLEKHL HSARSEEGRL YYDGEWYIRG QTICIDKKDE CPTSAVITTI NHDEVWFKRP DGSKSK Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 100%.
The protein encoded by this gene shows sequence similarity to the human breast carcinoma metastasis suppressor (BRMS1) protein and the mammalian Sds3 (suppressor of defective silencing 3) proteins. This protein is a component of the mSin3a family of histone deacetylase complexes (HDAC).
Specifications
| BRMS1L | |
| Polyclonal | |
| Unconjugated | |
| BRMS1L | |
| 0710008O11Rik; AI159718; breast cancer metastasis-suppressor 1; breast cancer metastasis-suppressor 1-like; breast cancer metastasis-suppressor 1-like protein; BRMS1; BRMS1-homolog protein p40; BRMS1L; BRMS1-like protein p40; D12Ertd407e | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84312 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5PSV4 | |
| BRMS1L | |
| Recombinant protein corresponding to Human BRMS1L. Recombinant protein control fragment (Product #RP-95570). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction