missing translation for 'onlineSavingsMsg'
Learn More

BRM Antibody, Novus Biologicals™

Product Code. 18341986 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341986 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341986 Supplier Novus Biologicals Supplier No. NBP313283

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BRM Polyclonal antibody specifically detects BRM in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRM
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 20% Glycerol (pH7)
Gene Alias ATP-dependent helicase SMARCA2, BAF190B, BRG1-associated factor 190B, BRMFLJ36757, EC 3.6.1, EC 3.6.4.-, global transcription activator homologous sequence, hBRMMGC74511, hSNF2a, probable global transcription activator SNF2L2, Protein brahma homolog, SNF2, SNF2/SWI2-like protein 2, SNF2A, SNF2-alpha, SNF2L2BAF190, SNF2LA, SNF2-like 2, Sth1p, subfamily a, member 2, sucrose nonfermenting 2-like protein 2, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin a2, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 2, SWI2
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human BRM. The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6595
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.