missing translation for 'onlineSavingsMsg'
Learn More

BRM Antibody (2D12), Novus Biologicals™

Product Code. 18326149 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326149 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18326149 Supplier Novus Biologicals Supplier No. H00006595M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BRM Monoclonal antibody specifically detects BRM in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRM
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 2D12
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_003061
Gene Alias ATP-dependent helicase SMARCA2, BAF190B, BRG1-associated factor 190B, BRMFLJ36757, EC 3.6.1, EC 3.6.4.-, global transcription activator homologous sequence, hBRMMGC74511, hSNF2a, probable global transcription activator SNF2L2, Protein brahma homolog, SNF2, SNF2/SWI2-like protein 2, SNF2A, SNF2-alpha, SNF2L2BAF190, SNF2LA, SNF2-like 2, Sth1p, subfamily a, member 2, sucrose nonfermenting 2-like protein 2, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin a2, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 2, SWI2
Host Species Mouse
Immunogen SMARCA2 (NP_003061, 659 a.a. ~ 735 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KILLDPNSEEVSEKDAKQIIETAKQDVDDEYSMQYSARGSQSYYTVAHAISERVEKQSALLINGTLKHYQLQGLEWM
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6595
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.